Assay Details
Gene Symbol: mgl-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083266.5 | NP_001076735.3 | 17 - 18 | 2816 | 110 | |
NM_001313057.3 | NP_001299986.1 | 11 - 12 | 1516 | 110 | ||
NM_001313058.3 | NP_001299987.1 | 11 - 12 | 1510 | 110 | ||
NM_001313059.3 | NP_001299988.1 | 7 - 8 | 922 | 110 | ||
NM_001313060.3 | NP_001299989.1 | 7 - 8 | 916 | 110 | ||
NM_171341.5 | NP_741400.2 | 17 - 18 | 2808 | 110 | ||
NR_132461.1 | 17 - 18 | 2888 | 110 | |||
NR_132462.1 | 16 - 17 | 2666 | 110 | |||
NR_132463.1 | 16 - 17 | 2657 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
