Assay Details
| Gene Symbol: mgl-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083266.5 | NP_001076735.3 | 1 - 2 | 391 | 107 | |
| NM_171341.5 | NP_741400.2 | 1 - 2 | 389 | 107 | ||
| NR_132461.1 | 1 - 2 | 321 | 107 | |||
| NR_132462.1 | 1 - 2 | 321 | 107 | |||
| NR_132463.1 | 1 - 2 | 321 | 107 | |||
More Information
Set Membership: |
|

mgl-3
Genomic Map