Assay Details
Gene Symbol: mgl-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083266.5 | NP_001076735.3 | 7 - 8 | 1416 | 96 | |
NM_001313057.3 | NP_001299986.1 | 1 - 2 | 116 | 96 | ||
NM_001313058.3 | NP_001299987.1 | 1 - 2 | 116 | 96 | ||
NM_171341.5 | NP_741400.2 | 7 - 8 | 1414 | 96 | ||
NR_132461.1 | 7 - 8 | 1346 | 96 | |||
NR_132462.1 | 7 - 8 | 1346 | 96 | |||
NR_132463.1 | 7 - 8 | 1346 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
