Assay Details
Gene Symbol: drp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028204.4 | NP_001023375.1 | 2 - 3 | 885 | 129 | |
NM_171344.5 | NP_741403.2 | 3 - 4 | 1039 | 129 | ||
GenBank mRNA | AF166274.1 | - | 2 - 3 | 892 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
