Assay Details
Gene Symbol: drp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028204.4 | NP_001023375.1 | 4 - 5 | 1755 | 107 | |
NM_171344.5 | NP_741403.2 | 5 - 6 | 1930 | 107 | ||
GenBank mRNA | AF166274.1 | - | 4 - 5 | 1762 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
