Assay Details
Gene Symbol: drp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028204.4 | NP_001023375.1 | 6 - 7 | 1939 | 65 | |
NM_171344.5 | NP_741403.2 | 7 - 8 | 2114 | 65 | ||
GenBank mRNA | AF166274.1 | - | 6 - 7 | 1946 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
