Assay Details
Gene Symbol: chn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059380.8 | NP_491781.2 | 2 - 3 | 131 | 102 | |
GenBank mRNA | AF303269.1 | - | 1 - 2 | 76 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
