Assay Details
| Gene Symbol: mut-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001372960.2 | NP_001360009.1 | 4 - 5 | 740 | 83 | |
| NM_059433.8 | NP_491834.1 | 4 - 5 | 739 | 83 | ||
More Information
Set Membership: |
|

mut-2
Genomic Map