Assay Details
| Gene Symbol: nars-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026234.7 | NP_001021405.1 | 2 - 3 | 173 | 101 | |
| NR_002344.1 | 1 - 2 | 82 | 101 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nars-1| Gene Symbol: nars-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026234.7 | NP_001021405.1 | 2 - 3 | 173 | 101 | |
| NR_002344.1 | 1 - 2 | 82 | 101 | |||
Set Membership: |
|