Assay Details
| Gene Symbol: adsl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380665.2 | NP_001367034.1 | 2 - 3 | 1321 | 68 | |
| NM_059648.6 | NP_492049.1 | 2 - 3 | 1302 | 68 | ||
More Information
Set Membership: |
|

adsl-1
Genomic Map