Assay Details
| Gene Symbol: unc-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026700.2 | NP_001021871.1 | 28 - 29 | 5342 | 78 | |
| NM_001026702.3 | NP_001021873.1 | 30 - 31 | 6368 | 78 | ||
| NM_001026703.4 | NP_001021874.1 | 29 - 30 | 5351 | 78 | ||
| NM_001263573.3 | NP_001250502.1 | 29 - 30 | 5360 | 78 | ||
| NM_001306224.3 | NP_001293153.1 | 17 - 18 | 4440 | 78 | ||
| NM_001330927.3 | NP_001317864.1 | 28 - 29 | 5351 | 78 | ||
| NM_001330928.3 | NP_001317865.1 | 28 - 29 | 5306 | 78 | ||
| NM_001330929.3 | NP_001317866.1 | 28 - 29 | 5297 | 78 | ||
| NM_001330930.3 | NP_001317863.1 | 20 - 21 | 4319 | 78 | ||
| NM_001330931.3 | NP_001317867.1 | 16 - 17 | 4298 | 78 | ||
| NM_001380703.1 | NP_001367037.1 | 21 - 22 | 4328 | 78 | ||
More Information
Set Membership: |
|

unc-13
Genomic Map