Assay Details
| Gene Symbol: pck-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380767.2 | NP_001366912.1 | 3 - 4 | 1579 | 76 | |
| NM_001380768.1 | NP_001367091.1 | 2 - 3 | 1202 | 76 | ||
| NM_001380772.1 | NP_001367090.1 | 1 - 2 | 1109 | 76 | ||
| NM_001383410.1 | 6 - 7 | 2826 | 76 | |||
More Information
Set Membership: |
|

pck-2
Genomic Map