Assay Details
| Gene Symbol: pck-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380767.2 | NP_001366912.1 | 4 - 5 | 1882 | 142 | |
| NM_001380768.1 | NP_001367091.1 | 3 - 4 | 1505 | 142 | ||
| NM_001380772.1 | NP_001367090.1 | 2 - 3 | 1412 | 142 | ||
| NM_001383410.1 | 7 - 8 | 3129 | 142 | |||
More Information
Set Membership: |
|

pck-2
Genomic Map