Assay Details
| Gene Symbol: che-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059820.4 | NP_492221.2 | 45 - 46 | 11551 | 125 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

che-3| Gene Symbol: che-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059820.4 | NP_492221.2 | 45 - 46 | 11551 | 125 | |
Set Membership: |
|