Assay Details
Gene Symbol: kri-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026692.5 | NP_001021863.1 | 6 - 7 | 965 | 85 | |
NM_001026693.5 | NP_001021864.1 | 6 - 7 | 960 | 85 | ||
GenBank mRNA | DQ372920.1 | - | 6 - 7 | 960 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
