Assay Details
Gene Symbol: gon-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129034.3 | NP_001122506.1 | 23 - 24 | 4824 | 119 | |
NM_001136316.4 | NP_001129788.1 | 17 - 18 | 3789 | 119 | ||
NM_001373178.2 | NP_001359707.1 | 25 - 26 | 4975 | 119 | ||
NM_059914.7 | NP_492315.4 | 26 - 27 | 5048 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
