Assay Details
| Gene Symbol: lmd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129003.3 | NP_001122475.1 | 1 - 2 | 190 | 95 | |
| NM_001129004.3 | NP_001122476.1 | 1 - 2 | 190 | 95 | ||
| Gene Symbol: F43G9.13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059928.5 | NP_492329.1 | 6 - 7 | 1783 | 95 | |
More Information
Set Membership: |
|

lmd-1
Genomic Map