Assay Details
Gene Symbol: cpn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383488.1 | 3 - 4 | 414 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: cpn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383488.1 | 3 - 4 | 414 | 66 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |