Assay Details
| Gene Symbol: cle-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059957.5 | NP_492358.3 | 14 - 15 | 1833 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cle-1| Gene Symbol: cle-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059957.5 | NP_492358.3 | 14 - 15 | 1833 | 124 | |
Set Membership: |
|