Assay Details
Gene Symbol: prp-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059962.4 | NP_492363.1 | 7 - 8 | 1180 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
