Assay Details
| Gene Symbol: gls-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059964.4 | NP_492365.3 | 14 - 15 | 2562 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gls-1| Gene Symbol: gls-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059964.4 | NP_492365.3 | 14 - 15 | 2562 | 91 | |
Set Membership: |
|