Assay Details
Gene Symbol: imp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025852.5 | NP_001021023.1 | 8 - 9 | 1458 | 133 | |
NM_001025853.5 | NP_001021024.1 | 8 - 9 | 1458 | 133 | ||
NM_001025854.6 | NP_001021025.1 | 8 - 9 | 1465 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
