Assay Details
| Gene Symbol: tin-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059969.6 | NP_492370.1 | 1 - 2 | 125 | 77 | |
| GenBank mRNA | AF144704.1 | - | 1 - 2 | 116 | 77 | |
More Information
Set Membership: |
|

tin-13
Genomic Map