Assay Details
| Gene Symbol: spd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060013.7 | NP_492414.1 | 6 - 7 | 1944 | 89 | |
| GenBank mRNA | AY340594.1 | - | 6 - 7 | 1895 | 89 | |
More Information
Set Membership: |
|

spd-2
Genomic Map