Assay Details
| Gene Symbol: gld-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060045.6 | NP_492446.3 | 5 - 6 | 780 | 76 | |
| GenBank mRNA | FJ610054.1 | - | 6 - 7 | 802 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gld-4| Gene Symbol: gld-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060045.6 | NP_492446.3 | 5 - 6 | 780 | 76 | |
| GenBank mRNA | FJ610054.1 | - | 6 - 7 | 802 | 76 | |
Set Membership: |
|