Assay Details
Gene Symbol: tat-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026286.5 | NP_001021457.1 | 5 - 6 | 1529 | 64 | |
NM_001026287.6 | NP_001021458.2 | 6 - 7 | 1436 | 64 | ||
NM_001047200.3 | NP_001040665.1 | 5 - 6 | 1520 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
