Assay Details
Gene Symbol: tat-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026286.5 | NP_001021457.1 | 6 - 7 | 2123 | 73 | |
NM_001026287.6 | NP_001021458.2 | 7 - 8 | 2030 | 73 | ||
NM_001047200.3 | NP_001040665.1 | 6 - 7 | 2114 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
