Assay Details
| Gene Symbol: rpoa-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060075.6 | NP_492476.1 | 8 - 9 | 2460 | 144 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rpoa-2| Gene Symbol: rpoa-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060075.6 | NP_492476.1 | 8 - 9 | 2460 | 144 | |
Set Membership: |
|