Assay Details
| Gene Symbol: pabp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060103.7 | NP_492504.1 | 1 - 2 | 97 | 69 | |
| Gene Symbol: C17E4.11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025830.7 | NP_001021001.1 | 1 - 2 | 91 | 69 | |
| Gene Symbol: marc-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060101.4 | NP_492502.2 | 1 - 2 | 93 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001025830.7, NM_060101.4 transcripts.
Set Membership: |
|

pabp-2
Genomic Map