Assay Details
Gene Symbol: ahr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025865.4 | NP_001021036.1 | 11 - 12 | 1646 | 70 | |
NM_001025866.5 | NP_001021037.1 | 10 - 11 | 1580 | 70 | ||
GenBank mRNA | AF039570.1 | - | 11 - 12 | 1642 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
