Assay Details
Gene Symbol: ahr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025865.4 | NP_001021036.1 | 2 - 3 | 123 | 66 | |
NM_001025866.5 | NP_001021037.1 | 1 - 2 | 57 | 66 | ||
GenBank mRNA | AF039570.1 | - | 2 - 3 | 119 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001025866.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
