Assay Details
Gene Symbol: ahr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025865.4 | NP_001021036.1 | 5 - 6 | 725 | 69 | |
NM_001025866.5 | NP_001021037.1 | 4 - 5 | 659 | 69 | ||
GenBank mRNA | AF039570.1 | - | 5 - 6 | 721 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
