Assay Details
Gene Symbol: ahr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025865.4 | NP_001021036.1 | 7 - 8 | 933 | 122 | |
NM_001025866.5 | NP_001021037.1 | 6 - 7 | 867 | 122 | ||
GenBank mRNA | AF039570.1 | - | 7 - 8 | 929 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
