Assay Details
Gene Symbol: ikb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060174.6 | NP_492575.1 | 3 - 4 | 436 | 82 | |
GenBank mRNA | AF348169.1 | - | 3 - 4 | 280 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
