Assay Details
| Gene Symbol: ikb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060174.6 | NP_492575.1 | 8 - 9 | 1519 | 81 | |
| GenBank mRNA | AF348169.1 | - | 8 - 9 | 1363 | 81 | |
More Information
Set Membership: |
|

ikb-1
Genomic Map