Assay Details
| Gene Symbol: mut-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060259.6 | NP_492660.3 | 1 - 2 | 242 | 70 | |
| NM_060260.9 | NP_492661.3 | 1 - 2 | 242 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mut-16| Gene Symbol: mut-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060259.6 | NP_492660.3 | 1 - 2 | 242 | 70 | |
| NM_060260.9 | NP_492661.3 | 1 - 2 | 242 | 70 | ||
Set Membership: |
|