Assay Details
| Gene Symbol: mut-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060259.6 | NP_492660.3 | 8 - 9 | 1735 | 96 | |
| NM_060260.9 | NP_492661.3 | 8 - 9 | 1723 | 96 | ||
More Information
Set Membership: |
|

mut-16
Genomic Map