Assay Details
| Gene Symbol: wts-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060298.5 | NP_492699.1 | 1 - 2 | 208 | 69 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length less than or equal to 70  Recommended Assay  Caenorhabditis elegans  Probe spans exons | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 wts-1
wts-1 Genomic Map
Genomic Map Back To Top
 Back To Top