Assay Details
| Gene Symbol: coa-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381194.1 | NP_001367155.1 | 4 - 5 | 447 | 111 | |
| NM_001381195.2 | NP_001366833.1 | 1 - 2 | 187 | 111 | ||
More Information
Set Membership: |
|

coa-6
Genomic Map