Assay Details
Gene Symbol: F25H2.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060358.4 | NP_492759.3 | 3 - 4 | 298 | 92 |
Gene Symbol: F25H2.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381201.1 | NP_001366855.1 | 3 - 4 | 317 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001381201.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
