Assay Details
| Gene Symbol: atm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306285.2 | NP_001293214.1 | 23 - 24 | 6083 | 110 | |
| NM_001306286.3 | NP_001293215.1 | 14 - 15 | 4040 | 110 | ||
More Information
Set Membership: |
|

atm-1
Genomic Map