Assay Details
| Gene Symbol: atm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306285.2 | NP_001293214.1 | 27 - 28 | 6902 | 90 | |
| NM_001306286.3 | NP_001293215.1 | 18 - 19 | 4859 | 90 | ||
More Information
Set Membership: |
|

atm-1
Genomic Map