Assay Details
| Gene Symbol: hum-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261950.4 | NP_001248879.1 | 1 - 2 | 210 | 89 | |
| NM_001261951.3 | NP_001248880.1 | 1 - 2 | 210 | 89 | ||
More Information
Set Membership: |
|

hum-7
Genomic Map