Assay Details
| Gene Symbol: pif-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058373.6 | NP_490774.2 | 2 - 3 | 464 | 69 | |
| GenBank mRNA | AB015041.1 | - | 2 - 3 | 321 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pif-1| Gene Symbol: pif-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058373.6 | NP_490774.2 | 2 - 3 | 464 | 69 | |
| GenBank mRNA | AB015041.1 | - | 2 - 3 | 321 | 69 | |
Set Membership: |
|