Assay Details
| Gene Symbol: pif-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306245.3 | NP_001293174.1 | 4 - 5 | 463 | 81 | |
| NM_058373.6 | NP_490774.2 | 6 - 7 | 930 | 81 | ||
| GenBank mRNA | AB015041.1 | - | 6 - 7 | 787 | 81 | |
More Information
Set Membership: |
|

pif-1
Genomic Map