Assay Details
Gene Symbol: shc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058398.5 | NP_490799.2 | 1 - 2 | 142 | 108 | |
NR_132554.1 | 1 - 2 | 3559 | 108 | |||
NR_132615.1 | 1 - 2 | 61 | 108 | |||
NR_132616.1 | 1 - 2 | 61 | 108 |
Gene Symbol: cutl-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058397.7 | NP_490798.1 | 1 - 2 | 142 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_058397.7 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
