Assay Details
Gene Symbol: shc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383442.1 | 4 - 5 | 763 | 70 | ||
NM_058398.5 | NP_490799.2 | 4 - 5 | 785 | 70 | ||
NR_132552.1 | 2 - 3 | 564 | 70 | |||
NR_132553.1 | 2 - 3 | 700 | 70 | |||
NR_132554.1 | 4 - 5 | 4338 | 70 | |||
NR_132615.1 | 4 - 5 | 840 | 70 | |||
NR_132616.1 | 3 - 4 | 850 | 70 |
Gene Symbol: cutl-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058397.7 | NP_490798.1 | 4 - 5 | 785 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_058397.7 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
