Assay Details
Gene Symbol: smo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058441.6 | NP_490842.1 | 2 - 3 | 190 | 89 | |
GenBank mRNA | X99600.1 | - | 165 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
