Assay Details
Gene Symbol: pcyt-2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129065.2 | NP_001122537.1 | 2 - 3 | 169 | 109 | |
NM_001306419.2 | NP_001293348.1 | 2 - 3 | 165 | 109 | ||
NM_058530.5 | NP_490931.1 | 2 - 3 | 167 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
