Assay Details
Gene Symbol: pcyt-2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306414.3 | NP_001293343.1 | 2 - 3 | 306 | 106 | |
NM_058530.5 | NP_490931.1 | 4 - 5 | 521 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
